Assay Details
| Gene Symbol: DAOA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040247.1 | 6 | 828 | 95 | ||
| GenBank mRNA | AY138548.1 | - | 6 | 828 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DAOA-AS1| Gene Symbol: DAOA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040247.1 | 6 | 828 | 95 | ||
| GenBank mRNA | AY138548.1 | - | 6 | 828 | 95 | |
Set Membership: |
|