Assay Details
| Gene Symbol: COL4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025912.1 | - | 16 | 100 | ||
| AW298730.1 | - | 1 | 316 | 100 | ||
| BC080644.1 | - | 6 | 100 | |||
| BX648393.1 | - | 17 | 100 | |||
| X05610.1 | - | 8 | 100 | |||
More Information
Set Membership: |
|

COL4A2
Genomic Map