Assay Details
| Gene Symbol: PRKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011536964.2 | XP_011535266.1 | 1 | 2710 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011536964.2 transcripts.
Set Membership: |
|

PRKD1
Genomic Map