Assay Details
| Gene Symbol: MIPOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020991.1 | XP_016876480.1 | 17 | 2693 | 105 | |
| XM_017020992.1 | XP_016876481.1 | 16 | 2556 | 105 | ||
| XM_017020993.1 | XP_016876482.1 | 16 | 2566 | 105 | ||
| XM_017020994.1 | XP_016876483.1 | 13 | 2211 | 105 | ||
| XM_017020995.1 | XP_016876484.1 | 15 | 2427 | 105 | ||
| XM_017020996.1 | XP_016876485.1 | 15 | 2443 | 105 | ||
| XM_017020997.1 | XP_016876486.1 | 14 | 2316 | 105 | ||
More Information
Set Membership: |
|

MIPOL1
Genomic Map