Assay Details
| Gene Symbol: ZNF106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284306.1 | NP_001271235.1 | 18 | 5921 | 113 | |
| NM_001284307.1 | NP_001271236.1 | 20 | 5936 | 113 | ||
| NM_022473.2 | NP_071918.1 | 19 | 8364 | 113 | ||
| XM_005254591.2 | XP_005254648.1 | 22 | 8271 | 113 | ||
| GenBank mRNA | AF205632.1 | - | 19 | 8337 | 113 | |
| AK055517.1 | - | 1 | 539 | 113 | ||
More Information
Set Membership: |
|

ZNF106
Genomic Map