Assay Details
| Gene Symbol: NPIPB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017022818.1 | XP_016878307.1 | 1 | 330 | 98 | |
| XM_017022822.1 | XP_016878311.1 | 1 | 328 | 98 | ||
| XM_017022823.1 | XP_016878312.1 | 1 | 329 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017022818.1, XM_017022822.1, XM_017022823.1 transcripts.
Set Membership: |
|

NPIPB9
Genomic Map