Assay Details
| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002196.2 | 1 | 360 | 61 | ||
| NR_131223.1 | 1 | 360 | 61 | |||
| GenBank mRNA | AL546946.3 | - | 1 | 320 | 61 | |
| AL548405.3 | - | 1 | 320 | 61 | ||
| DA443982.1 | - | 1 | 360 | 61 | ||
| Gene Symbol: RPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC002550.1 | - | 150 | 61 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AF118081.1 | - | Chr. - - [-] on Build | 61 | |||
More Information
Set Membership: |
|

H19
Genomic Map