Assay Details
| Gene Symbol: YWHAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135699.1 | NP_001129171.1 | 6 | 2917 | 104 | |
| NM_001135700.1 | NP_001129172.1 | 6 | 2871 | 104 | ||
| NM_001135701.1 | NP_001129173.1 | 6 | 2920 | 104 | ||
| NM_001135702.1 | NP_001129174.1 | 6 | 2939 | 104 | ||
| NM_003406.3 | NP_003397.1 | 6 | 2900 | 104 | ||
| NM_145690.2 | NP_663723.1 | 6 | 2974 | 104 | ||
| XM_005251061.3 | XP_005251118.1 | 8 | 3297 | 104 | ||
| XM_005251063.3 | XP_005251120.1 | 6 | 3069 | 104 | ||
| XM_017013810.1 | XP_016869299.1 | 7 | 3086 | 104 | ||
| XM_017013811.1 | XP_016869300.1 | 8 | 3282 | 104 | ||
| GenBank mRNA | BM783874.1 | - | 1 | 234 | 104 | |
More Information
Set Membership: |
|

YWHAZ
Genomic Map