Assay Details
| Gene Symbol: LRRC37BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124001.1 | - | 2 | 2015 | 101 | |
| AK124759.1 | - | 2 | 2422 | 101 | ||
| AL713754.1 | - | 2 | 1540 | 101 | ||
| Gene Symbol: LOC107133515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015341.2 | 9 | 2277 | 101 | ||
| GenBank mRNA | AK075282.1 | - | 6 | 1553 | 101 | |
| Gene Symbol: LRRC37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123784.1 | - | 1 | 126 | 101 | |
More Information
Set Membership: |
|

LRRC37BP1
Genomic Map