Assay Details
| Gene Symbol: TBC1D3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006722235.2 | XP_006722298.1 | 1 | 104 | 113 | |
| XM_006722236.1 | XP_006722299.1 | 1 | 104 | 113 | ||
| XM_006722237.2 | XP_006722300.1 | 1 | 104 | 113 | ||
| XM_006722239.2 | XP_006722302.1 | 1 | 104 | 113 | ||
| XM_011524172.1 | XP_011522474.1 | 1 | 104 | 113 | ||
| XM_011524173.1 | XP_011522475.1 | 1 | 104 | 113 | ||
| XM_011524174.1 | XP_011522476.1 | 1 | 104 | 113 | ||
| Gene Symbol: TBC1D3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005276914.3 | XP_005276971.1 | 1 | 1079 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005276914.3, XM_006722235.2, XM_006722236.1, XM_006722237.2, XM_006722239.2, XM_011524172.1, XM_011524173.1, XM_011524174.1 transcripts.
Set Membership: |
|

TBC1D3K
Genomic Map