Assay Details
| Gene Symbol: ITPKB-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103784.1 | 2 | 1228 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPKB-IT1| Gene Symbol: ITPKB-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103784.1 | 2 | 1228 | 104 | ||
Set Membership: |
|