Assay Details
| Gene Symbol: CCDC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316321.1 | NP_001303250.1 | 15 | 4699 | 70 | |
| GenBank mRNA | AK074059.1 | - | 7 | 3158 | 70 | |
| AK125941.1 | - | 1 | 1293 | 70 | ||
More Information
Set Membership: |
|

CCDC57
Genomic Map