Assay Details
| Gene Symbol: TIMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013337.2 | NP_037469.2 | 4 | 875 | 113 | |
| GenBank mRNA | AF155330.1 | - | 4 | 854 | 113 | |
| AK000830.1 | - | 4 | 856 | 113 | ||
| BC002324.2 | - | 4 | 875 | 2819655 | 113 | |
More Information
Set Membership: |
|

TIMM22
Genomic Map