Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011523902.2 | XP_011522204.1 | 1 | 66 | 111 | |
| XM_017024700.1 | XP_016880189.1 | 1 | 66 | 111 | ||
| XM_017024703.1 | XP_016880192.1 | 1 | 612 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011523902.2, XM_017024700.1 transcripts.
Set Membership: |
|

PAFAH1B1
Genomic Map