Assay Details
| Gene Symbol: SH3GL1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033412.1 | 1 | 1811 | 99 | ||
| Gene Symbol: SH3GL1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033420.1 | 1 | 1800 | 99 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30846127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16967902 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3564394 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3564399 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25006 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 55984 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3564399 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3545065 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3564394 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3564399 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3564394 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1567027_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1567028_PM_s_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1567030_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004420.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004420.hg.1 | TC17000353.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC17000353.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153396.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153396.hg.1 | TC1700007461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700153396.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153397.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153397.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153397.hg.1 | TC1700007461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700153397.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153397.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007461.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007461.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153399.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153399.hg.1 | TC1700007461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700153399.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153398.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153398.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153398.hg.1 | TC1700007461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700153398.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153398.hg.1 | NR_033420.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1567028_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567030_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567028_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567030_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567027_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567027_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567028_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567030_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567027_at10 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009342 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009372 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009372 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009342 | - | - |

SH3GL1P1
Genomic Map