Assay Details
| Gene Symbol: RAB11FIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303542.2 | NP_001290471.2 | 13 | 69 | ||
| NM_032932.5 | NP_116321.2 | 15 | 69 | |||
| GenBank mRNA | AB058724.1 | - | 15 | 69 | ||
| AJ314646.2 | - | 15 | 69 | |||
| AK055019.1 | - | 1 | 69 | |||
| AY169244.1 | - | 15 | 69 | |||
| BC131530.1 | - | 1 | 725 | 69 | ||
More Information
Set Membership: |
|

RAB11FIP4
Genomic Map