Assay Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011526568.2 | XP_011524870.1 | 1 | 235 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011526568.2 transcripts.
Set Membership: |
|

ZNF550
Genomic Map