Assay Details
| Gene Symbol: LILRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011526331.2 | XP_011524633.1 | 14 | 2784 | 193 | |
| XM_011526332.2 | XP_011524634.1 | 14 | 2781 | 193 | ||
| XM_011526335.2 | XP_011524637.1 | 13 | 2625 | 193 | ||
| XM_011526336.2 | XP_011524638.1 | 13 | 2592 | 193 | ||
| XM_017026182.1 | XP_016881671.1 | 14 | 2781 | 193 | ||
| XM_017026183.1 | XP_016881672.1 | 14 | 2778 | 193 | ||
| XM_017026184.1 | XP_016881673.1 | 14 | 2778 | 193 | ||
| XM_017026185.1 | XP_016881674.1 | 14 | 2747 | 193 | ||
| XM_017026188.1 | XP_016881677.1 | 15 | 2903 | 193 | ||
| XM_017026189.1 | XP_016881678.1 | 15 | 2903 | 193 | ||
| XM_017026190.1 | XP_016881679.1 | 16 | 3255 | 193 | ||
| XM_017026191.1 | XP_016881680.1 | 13 | 2595 | 193 | ||
More Information
Set Membership: |
|

LILRB1
Genomic Map