Assay Details
| Gene Symbol: LILRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011526331.2 | XP_011524633.1 | 14 | 2686 | 157 | |
| XM_011526332.2 | XP_011524634.1 | 14 | 2683 | 157 | ||
| XM_011526335.2 | XP_011524637.1 | 13 | 2527 | 157 | ||
| XM_011526336.2 | XP_011524638.1 | 13 | 2494 | 157 | ||
| XM_017026182.1 | XP_016881671.1 | 14 | 2683 | 157 | ||
| XM_017026183.1 | XP_016881672.1 | 14 | 2680 | 157 | ||
| XM_017026184.1 | XP_016881673.1 | 14 | 2680 | 157 | ||
| XM_017026185.1 | XP_016881674.1 | 14 | 2649 | 157 | ||
| XM_017026188.1 | XP_016881677.1 | 15 | 2805 | 157 | ||
| XM_017026189.1 | XP_016881678.1 | 15 | 2805 | 157 | ||
| XM_017026190.1 | XP_016881679.1 | 16 | 3157 | 157 | ||
| XM_017026191.1 | XP_016881680.1 | 13 | 2497 | 157 | ||
More Information
Set Membership: |
|

LILRB1
Genomic Map