Assay Details
| Gene Symbol: TRIM56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011516589.2 | XP_011514891.1 | 2 | 5962 | 78 | |
| Gene Symbol: C11orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130935.1 | 3 | 1210 | 78 | ||
| Gene Symbol: FKSG49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF336882.1 | - | 1 | 78 | ||
| AF336883.1 | - | 1 | 78 | |||
| AF338190.1 | - | 1 | 78 | |||
| AF338193.1 | - | 1 | 78 | |||
| AF338194.1 | - | 1 | 78 | |||
| AF338195.1 | - | 1 | 78 | |||
| AF469044.1 | - | 1 | 78 | |||
| Gene Symbol: HELLPAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JX088243.1 | - | 1 | 78 | ||
| Gene Symbol: LOC100506634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF469043.1 | - | 2 | 2656 | 78 | |
| Gene Symbol: NEK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126330.1 | - | 9 | 78 | ||
| Gene Symbol: SLC38A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128743.1 | - | 2929 | 78 | ||
| Gene Symbol: TDRKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK225160.1 | - | 15 | 3492 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF336882.1, AF336883.1, AF338190.1, AF338193.1, AF338194.1, AF338195.1, AF469044.1 transcripts.
Set Membership: |
|

TRIM56
Genomic Map