Assay Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017027873.1 | XP_016883362.1 | 1 - 2 | 140 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017027873.1 transcripts.
Set Membership: |
|

OSER1
Genomic Map