Assay Details
| Gene Symbol: GORASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006713301.3 | XP_006713364.1 | 1 | 404 | 59 | |
| XM_011534020.2 | XP_011532322.1 | 1 | 404 | 59 | ||
| XM_011534021.2 | XP_011532323.1 | 1 | 404 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006713301.3, XM_011534020.2, XM_011534021.2 transcripts.
Set Membership: |
|

GORASP1
Genomic Map