Assay Details
| Gene Symbol: RALBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006788.3 | NP_006779.1 | 10 | 3650 | 77 | |
| GenBank mRNA | AY517554.1 | - | 8 | 2775 | 77 | |
| BC013126.1 | - | 10 | 3512 | 3049077 | 77 | |
| L42542.1 | - | 10 | 3658 | 77 | ||
More Information
Set Membership: |
|

RALBP1
Genomic Map