Assay Details
| Gene Symbol: HTT-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045414.1 | 3 | 398 | 101 | ||
| GenBank mRNA | DA153759.1 | - | 3 | 398 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTT-AS| Gene Symbol: HTT-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045414.1 | 3 | 398 | 101 | ||
| GenBank mRNA | DA153759.1 | - | 3 | 398 | 101 | |
Set Membership: |
|