Assay Details
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF216184.1 | - | 24 | 157 | ||
| BM670090.1 | - | 1 | 240 | 157 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF216184.1 | - | 24 | 157 | ||
| BM670090.1 | - | 1 | 240 | 157 | ||
Set Membership: |
|