Assay Details
| Gene Symbol: MAPK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318067.1 | NP_001304996.1 | 13 | 4835 | 131 | |
| NM_001318068.1 | NP_001304997.1 | 13 | 4930 | 131 | ||
| NM_001318069.1 | NP_001304998.1 | 14 | 5400 | 131 | ||
| NM_002753.4 | NP_002744.1 | 14 | 4941 | 131 | ||
| NM_138980.3 | NP_620446.1 | 14 | 5427 | 131 | ||
| NM_138982.3 | NP_620448.1 | 14 | 5400 | 131 | ||
| XM_005263131.2 | XP_005263188.1 | 14 | 5294 | 131 | ||
More Information
Set Membership: |
|

MAPK10
Genomic Map