Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128426.2 | NP_001121898.1 | 9 | 3015 | 144 | |
| NM_018569.5 | NP_061039.3 | 10 | 3114 | 144 | ||
| NR_136622.1 | 10 | 3110 | 144 | |||
| GenBank mRNA | BX498932.1 | - | 1 | 144 | ||
More Information
Set Membership: |
|

AP1AR
Genomic Map