Assay Details
| Gene Symbol: ATF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005273146.1 | XP_005273203.1 | 1 | 286 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005273146.1 transcripts.
Set Membership: |
|

ATF3
Genomic Map