Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005268492.2 | XP_005268549.1 | 3 | 12858 | 124 | |
| XM_005268493.2 | XP_005268550.1 | 3 | 12254 | 124 | ||
| XM_006714789.2 | XP_006714852.1 | 3 | 12918 | 124 | ||
| XM_011537671.2 | XP_011535973.1 | 5 | 12753 | 124 | ||
| GenBank mRNA | AK091723.1 | - | 1 | 147 | 124 | |
| BC036491.1 | - | 1 | 1964 | 124 | ||
More Information
Set Membership: |
|

KCTD16
Genomic Map