Assay Details
| Gene Symbol: CCT6P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033416.1 | 11 | 1826 | 150 | ||
| GenBank mRNA | BX649060.1 | - | 9 | 3291 | 150 | |
| Gene Symbol: CCT6P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092180.1 | - | 2 | 1580 | 150 | |
More Information
Set Membership: |
|

CCT6P3
Genomic Map