Assay Details
| Gene Symbol: PHF20L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017013513.1 | XP_016869002.1 | 5 | 2040 | 87 | |
| XM_017013514.1 | XP_016869003.1 | 5 | 2036 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017013513.1, XM_017013514.1 transcripts.
Set Membership: |
|

PHF20L1
Genomic Map