Assay Details
| Gene Symbol: PXDNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006716420.3 | XP_006716483.1 | 1 | 869 | 71 | |
| XM_011517456.2 | XP_011515758.1 | 1 | 869 | 71 | ||
| XM_011517457.2 | XP_011515759.1 | 1 | 869 | 71 | ||
| XM_011517459.2 | XP_011515761.1 | 1 | 869 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006716420.3, XM_011517456.2, XM_011517457.2, XM_011517459.2 transcripts.
Set Membership: |
|

PXDNL
Genomic Map