Assay Details
| Gene Symbol: CDC40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015891.2 | NP_056975.1 | 15 | 2239 | 73 | |
| GenBank mRNA | AB208903.1 | - | 15 | 2214 | 73 | |
| AK001426.1 | - | 4 | 901 | 73 | ||
| BC117145.1 | - | 15 | 2236 | 40125696 | 73 | |
| BC126114.1 | - | 15 | 2236 | 8991830 | 73 | |
More Information
Set Membership: |
|

CDC40
Genomic Map