Assay Details
| Gene Symbol: VSTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017015828.1 | XP_016871317.1 | 1 | 296 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017015828.1 transcripts.
Set Membership: |
|

VSTM4
Genomic Map