Assay Details
| Gene Symbol: H1FNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181788.1 | NP_861453.1 | 1 | 1174 | 72 | |
| GenBank mRNA | AB121028.2 | - | 1 | 814 | 72 | |
| AY302593.1 | - | 1 | 1174 | 72 | ||
| BC118635.1 | - | 1 | 1122 | 40023772 | 72 | |
| BC119787.1 | - | 1 | 1124 | 40023773 | 72 | |
More Information
Set Membership: |
|

H1FNT
Genomic Map