Assay Details
| Gene Symbol: MAPKAPK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020127.1 | XP_016875616.1 | 15 | 3816 | 74 | |
| XM_017020128.1 | XP_016875617.1 | 15 | 2267 | 74 | ||
| XM_017020132.1 | XP_016875621.1 | 16 | 3864 | 74 | ||
| XM_017020133.1 | XP_016875622.1 | 15 | 3636 | 74 | ||
| XM_017020134.1 | XP_016875623.1 | 15 | 3462 | 74 | ||
| XM_017020135.1 | XP_016875624.1 | 14 | 3545 | 74 | ||
| XM_017020136.1 | XP_016875625.1 | 15 | 3630 | 74 | ||
| XM_017020138.1 | XP_016875627.1 | 11 | 3023 | 74 | ||
More Information
Set Membership: |
|

MAPKAPK5
Genomic Map