Assay Details
| Gene Symbol: P2RY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002564.3 | NP_002555.3 | 3 | 2814 | 109 | |
| NM_176071.2 | NP_788085.2 | 3 | 2879 | 109 | ||
| NM_176072.2 | NP_788086.2 | 3 | 2948 | 109 | ||
More Information
Set Membership: |
|

P2RY2
Genomic Map