Assay Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106.3 | NP_001097.2 | 11 | 4545 | 89 | |
| XM_005265583.3 | XP_005265640.1 | 11 | 7007 | 89 | ||
| XM_017007514.1 | XP_016863003.1 | 11 | 6986 | 89 | ||
| XM_017007515.1 | XP_016863004.1 | 11 | 4631 | 89 | ||
| XM_017007516.1 | XP_016863005.1 | 11 | 7955 | 89 | ||
More Information
Set Membership: |
|

ACVR2B
Genomic Map