Assay Details
| Gene Symbol: DCAF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248169.3 | XP_005248226.1 | 3 | 5346 | 87 | |
| XM_005248171.3 | XP_005248228.1 | 2 | 5242 | 87 | ||
| XM_006713966.3 | XP_006714029.1 | 4 | 5517 | 87 | ||
| XM_017008327.1 | XP_016863816.1 | 2 | 6811 | 87 | ||
| XM_017008328.1 | XP_016863817.1 | 2 | 6796 | 87 | ||
More Information
Set Membership: |
|

DCAF16
Genomic Map