Assay Details
| Gene Symbol: ANKRD33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164440.1 | NP_001157912.1 | 4 | 4211 | 90 | |
| XM_005248343.4 | XP_005248400.1 | 3 | 3656 | 90 | ||
| XM_005248344.3 | XP_005248401.1 | 3 | 3646 | 90 | ||
More Information
Set Membership: |
|

ANKRD33B
Genomic Map