Assay Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000112.3 | NP_000103.2 | 3 | 3977 | 94 | |
| XM_017009191.1 | XP_016864680.1 | 4 | 3882 | 94 | ||
| GenBank mRNA | AK025078.1 | - | 1 | 244 | 94 | |
| BX640696.1 | - | 2 | 1770 | 94 | ||
More Information
Set Membership: |
|

SLC26A2
Genomic Map