Assay Details
| Gene Symbol: FLNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127487.1 | NP_001120959.1 | 46 | 7995 | 93 | |
| NM_001458.4 | NP_001449.3 | 47 | 8094 | 93 | ||
| GenBank mRNA | AB208865.1 | - | 24 | 3736 | 93 | |
| AB371585.1 | - | 47 | 8114 | 93 | ||
| AF089841.1 | - | 47 | 7934 | 93 | ||
| AF146692.1 | - | 46 | 7995 | 93 | ||
| AJ012737.1 | - | 46 | 7804 | 93 | ||
| AK001048.1 | - | 3 | 404 | 93 | ||
| AK126540.1 | - | 1 | 2327 | 93 | ||
More Information
Set Membership: |
|

FLNC
Genomic Map