Assay Details
| Gene Symbol: SLC39A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128431.2 | NP_001121903.1 | 9 | 2915 | 119 | |
| NM_001135153.1 | NP_001128625.1 | 9 | 2956 | 119 | ||
| NM_015359.4 | NP_056174.2 | 9 | 2915 | 119 | ||
| XM_005273465.2 | XP_005273522.1 | 10 | 3003 | 119 | ||
| XM_005273466.4 | XP_005273523.1 | 9 | 2987 | 119 | ||
| XM_006716323.2 | XP_006716386.1 | 11 | 3201 | 119 | ||
| XM_006716324.2 | XP_006716387.1 | 10 | 2992 | 119 | ||
| XM_011544478.2 | XP_011542780.1 | 10 | 3067 | 119 | ||
| XM_017013292.1 | XP_016868781.1 | 10 | 3083 | 119 | ||
| XM_017013293.1 | XP_016868782.1 | 11 | 3201 | 119 | ||
| XM_017013294.1 | XP_016868783.1 | 11 | 3110 | 119 | ||
| XM_017013295.1 | XP_016868784.1 | 9 | 3120 | 119 | ||
| GenBank mRNA | AK172810.1 | - | 9 | 2885 | 118 | |
| BX648538.1 | - | 2 | 1269 | 118 | ||
| D31887.1 | - | 9 | 2859 | 119 | ||
More Information
Set Membership: |
|

SLC39A14
Genomic Map