Assay Details
| Gene Symbol: LAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122606.1 | NP_001116078.1 | 9 | 3639 | 86 | |
| NM_002294.2 | NP_002285.1 | 9 | 6475 | 86 | ||
| GenBank mRNA | BC040653.2 | - | 1 | 2005 | 86 | |
More Information
Set Membership: |
|

LAMP2
Genomic Map