Assay Details
| Gene Symbol: ARID5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244638.1 | NP_001231567.1 | 7 | 3335 | 64 | |
| NM_032199.2 | NP_115575.1 | 10 | 4251 | 64 | ||
| XM_011540262.1 | XP_011538564.1 | 9 | 4020 | 64 | ||
| XM_017016765.1 | XP_016872254.1 | 5 | 3009 | 64 | ||
| GenBank mRNA | AK123882.1 | - | 1 | 495 | 64 | |
| BC036831.1 | - | 1 | 2379 | 64 | ||
| BX352258.2 | - | 1 | 64 | |||
| BX537690.1 | - | 1 | 1758 | 64 | ||
| BX641020.1 | - | 1 | 2039 | 64 | ||
| BX648835.1 | - | 1 | 609 | 64 | ||
More Information
Set Membership: |
|

ARID5B
Genomic Map