Assay Details
| Gene Symbol: IPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270839.4 | XP_005270896.1 | 9 | 7060 | 115 | |
| XM_006710623.3 | XP_006710686.1 | 8 | 6951 | 115 | ||
More Information
Set Membership: |
|

IPP
Genomic Map