Assay Details
| Gene Symbol: IPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145349.1 | NP_001138821.1 | 5 | 1241 | 84 | |
| NM_005897.2 | NP_005888.1 | 5 | 1241 | 84 | ||
| XM_005270839.4 | XP_005270896.1 | 5 | 1241 | 84 | ||
| XM_006710623.3 | XP_006710686.1 | 5 | 1241 | 84 | ||
| XM_011541394.2 | XP_011539696.1 | 5 | 1241 | 84 | ||
| XM_017001210.1 | XP_016856699.1 | 5 | 1241 | 84 | ||
| GenBank mRNA | AF156857.1 | - | 4 | 1024 | 84 | |
| AK023880.1 | - | 5 | 1077 | 84 | ||
| BC032544.1 | - | 5 | 1082 | 5535760 | 84 | |
More Information
Set Membership: |
|

IPP
Genomic Map