Assay Details
| Gene Symbol: LRRC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534989.2 | XP_011533291.1 | 7 | 1753 | 68 | |
| XM_017020424.1 | XP_016875913.1 | 8 | 5126 | 68 | ||
More Information
Set Membership: |
|

LRRC63
Genomic Map