Assay Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002023.4 | NP_002014.2 | 3 | 1720 | 77 | |
| NR_103757.1 | 2 | 734 | 77 | |||
| GenBank mRNA | AK092899.1 | - | 4 | 1181 | 77 | |
| AK291632.1 | - | 3 | 1386 | 77 | ||
| BC035281.1 | - | 3 | 1397 | 5181410 | 77 | |
| U05291.1 | - | 2 | 363 | 77 | ||
More Information
Set Membership: |
|

FMOD
Genomic Map