Assay Details
| Gene Symbol: MAPK1IP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144578.3 | NP_653179.1 | 4 | 134 | ||
| XM_011537362.2 | XP_011535664.2 | 4 | 134 | |||
| GenBank mRNA | AK054650.1 | - | 1 | 134 | ||
More Information
Set Membership: |
|

MAPK1IP1L
Genomic Map